Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318859.1 | NP_001305788.1 | 4 - 5 | 611 | 127 |
NM_001318860.1 | NP_001305789.1 | 5 - 6 | 780 | 127 | |
NM_001318861.1 | NP_001305790.1 | 6 - 7 | 866 | 127 | |
NM_001318862.1 | NP_001305791.1 | 6 - 7 | 848 | 127 | |
NM_001318863.1 | NP_001305792.1 | 5 - 6 | 762 | 127 | |
NM_080916.2 | NP_550438.1 | 6 - 7 | 893 | 127 | |
NM_080918.2 | NP_550440.1 | 4 - 5 | 629 | 127 | |
NR_134893.1 | 3 - 4 | 516 | 127 | ||
NR_134894.1 | 4 - 5 | 664 | 127 | ||
NR_134895.1 | 2 - 3 | 328 | 127 | ||
NR_134896.1 | 3 - 4 | 498 | 127 | ||
NR_134897.1 | 5 - 6 | 708 | 127 | ||
NR_134898.1 | 4 - 5 | 632 | 127 | ||
XM_011532647.2 | XP_011530949.1 | 6 - 7 | 871 | 127 | |
GenBank mRNA | AI201630.1 | - | 1 - 2 | 127 | |
AV760728.1 | - | - | 261 | 127 | |
BC001121.2 | - | 3 - 4 | 455 | 127 | |
BC015757.1 | - | 6 - 7 | 832 | 127 | |
BC024019.1 | - | 3 - 4 | 470 | 127 | |
BM706344.1 | - | 3 - 4 | 429 | 127 | |
CA414889.1 | - | 1 - 2 | 127 | ||
CN311963.1 | - | 5 - 6 | 637 | 127 | |
U41668.1 | - | 6 - 7 | 842 | 127 |