Assay Design
Gene Transcripts
- Gene Symbol
- HIST1H2AM
- Entrez Gene ID
-
8336
- Gene Name
- histone cluster 1, H2am
- Gene Aliases
-
H2A.1, H2A/n, H2AFN, dJ193B12.1
- Chromosome Location
-
Chr.6: 27892699 - 27893185 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003514.2 |
NP_003505.1 |
1 - 1 |
52 |
82 |
GenBank mRNA |
AK312224.1 |
- |
1 - 1 |
52 |
82 |
View Full Product Details