Assay Design



Gene Transcripts

Gene Symbol
HBE1
Entrez Gene ID
3046
Gene Name
hemoglobin subunit epsilon 1
Gene Aliases
HBE
Chromosome Location
Chr.11: 5268350 - 5270143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005330.3 NP_005321.1 2 - 3 566 79
GenBank mRNA BC015537.2 - 4 - 5 658 79
CR541912.1 - 2 - 3 313 79

View Full Product Details