Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040118.2 | NP_001035207.1 | 27 - 28 | 3889 | 76 |
NM_001135190.1 | NP_001128662.1 | 24 - 25 | 3517 | 76 | |
NM_015242.4 | NP_056057.2 | 25 - 26 | 3700 | 76 | |
GenBank mRNA | AB018325.2 | - | 25 - 26 | 3677 | 76 |
AF411983.1 | - | 21 - 22 | 2758 | 76 | |
AJ621557.1 | - | 25 - 26 | 3700 | 76 | |
AK160383.1 | - | 23 - 24 | 4367 | 76 | |
AY049732.1 | - | 22 - 23 | 3537 | 76 | |
AY553630.1 | - | 27 - 28 | 3904 | 76 | |
BC008315.1 | - | 9 - 10 | 1170 | 76 | |
BC056401.1 | - | 13 - 14 | 1543 | 76 | |
BC140792.1 | - | 24 - 25 | 3187 | 76 | |
BC144562.1 | - | 25 - 26 | 3370 | 76 |