Assay Design



Gene Transcripts

Gene Symbol
USP28
Entrez Gene ID
57646
Gene Name
ubiquitin specific peptidase 28
Gene Aliases
-
Chromosome Location
Chr.11: 113797875 - 113875580 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301029.1 NP_001287958.1 14 - 15 1835 109
NM_020886.3 NP_065937.1 16 - 17 2046 109
XM_005271630.2 XP_005271687.1 16 - 17 2045 109
XM_005271631.2 XP_005271688.1 16 - 17 2042 109
XM_005271632.2 XP_005271689.1 17 - 18 2375 109
XM_005271633.4 XP_005271690.1 17 - 18 2103 109
XM_005271636.2 XP_005271693.1 15 - 16 1865 109
XM_005271637.2 XP_005271694.1 16 - 17 2045 109
XM_005271638.2 XP_005271695.1 16 - 17 2045 109
XM_005271639.2 XP_005271696.1 16 - 17 2042 109
XM_011542936.2 XP_011541238.1 15 - 16 1964 109
XM_011542938.1 XP_011541240.1 14 - 15 1838 109
XM_011542941.1 XP_011541243.1 9 - 10 1221 109
XM_011542942.1 XP_011541244.1 5 - 6 761 109
XM_017018056.1 XP_016873545.1 15 - 16 1961 109
XM_017018057.1 XP_016873546.1 17 - 18 2372 109
XM_017018058.1 XP_016873547.1 15 - 16 1978 109
XM_017018059.1 XP_016873548.1 15 - 16 1961 109
XM_017018060.1 XP_016873549.1 14 - 15 1835 109
XM_017018061.1 XP_016873550.1 14 - 15 1838 109
XM_017018062.1 XP_016873551.1 14 - 15 1835 109
XM_017018063.1 XP_016873552.1 15 - 16 1922 109
XM_017018064.1 XP_016873553.1 14 - 15 1843 109
XM_017018065.1 XP_016873554.1 17 - 18 2087 109
XM_017018066.1 XP_016873555.1 18 - 19 2449 109
XM_017018067.1 XP_016873556.1 13 - 14 1698 109
XM_017018068.1 XP_016873557.1 9 - 10 1221 109
XM_017018069.1 XP_016873558.1 5 - 6 761 109
GenBank mRNA AB040948.1 - 7 - 8 1189 109
AF266283.1 - 16 - 17 1983 109
AK097623.1 - 16 - 17 1676 109
AK304770.1 - 14 - 15 1806 109
BU940680.1 - 1 - 2 78 109

View Full Product Details