Assay Design



Gene Transcripts

Gene Symbol
PRAMEF1
Entrez Gene ID
65121
Gene Name
PRAME family member 1
Gene Aliases
-
Chromosome Location
Chr.1: 12791397 - 12796628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023013.4 NP_075389.2 3 - 4 969 133
GenBank mRNA AK125568.1 - 3 - 4 969 133
AL049686.1 - 3 - 4 969 133
BC144199.1 - 3 - 4 939 133

Gene Symbol
PRAMEF13
Entrez Gene ID
400736
Gene Name
PRAME family member 13
Gene Aliases
-
Chromosome Location
Chr.1: 13196188 - 13201409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291380.1 NP_001278309.1 3 - 4 969 133

Gene Symbol
PRAMEF14
Entrez Gene ID
729528
Gene Name
PRAME family member 14
Gene Aliases
-
Chromosome Location
Chr.1: 13341892 - 13347134 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024661.2 NP_001019832.2 3 - 4 969 133

Gene Symbol
PRAMEF2
Entrez Gene ID
65122
Gene Name
PRAME family member 2
Gene Aliases
DJ845O24.3
Chromosome Location
Chr.1: 12857086 - 12861909 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023014.1 NP_075390.1 3 - 4 953 133
GenBank mRNA AL049674.1 - 3 - 4 953 133
BC075008.2 - 2 - 3 902 133
BC075009.2 - 2 - 3 902 133

View Full Product Details