Assay Design
Gene Transcripts
- Gene Symbol
- SCRT1
- Entrez Gene ID
-
83482
- Gene Name
- scratch family transcriptional repressor 1
- Gene Aliases
-
SCRT, ZNF898
- Chromosome Location
-
Chr.8: 144330565 - 144336281 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_031309.5 |
NP_112599.2 |
1 - 2 |
221 |
116 |
GenBank mRNA |
AY014996.1 |
- |
1 - 2 |
221 |
116 |
View Full Product Details