Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243476.2 | NP_001230405.1 | 16 - 17 | 3519 | 65 |
NM_052851.2 | NP_443083.1 | 12 - 13 | 3109 | 65 | |
NM_178006.3 | NP_821074.1 | 12 - 13 | 3172 | 65 | |
NM_178007.2 | NP_821075.1 | 12 - 13 | 3079 | 65 | |
XM_011535299.2 | XP_011533601.1 | 15 - 16 | 3265 | 65 | |
XM_017020834.1 | XP_016876323.1 | 12 - 13 | 3277 | 65 | |
XM_017020835.1 | XP_016876324.1 | 14 - 15 | 3198 | 65 | |
GenBank mRNA | AK094704.1 | - | 11 - 12 | 2070 | 65 |
AK095114.1 | - | 8 - 9 | 1792 | 65 | |
AK312888.1 | - | 12 - 13 | 3165 | 65 | |
AL049801.1 | - | 12 - 13 | 3065 | 65 | |
AY082589.1 | - | 12 - 13 | 3167 | 65 | |
AY082590.1 | - | 12 - 13 | 3091 | 65 | |
AY082591.1 | - | 12 - 13 | 3065 | 65 | |
AY366448.1 | - | 12 - 13 | 3107 | 65 | |
BX647695.1 | - | 16 - 17 | 3479 | 65 |