Assay Design



Gene Transcripts

Gene Symbol
PGLYRP3
Entrez Gene ID
114771
Gene Name
peptidoglycan recognition protein 3
Gene Aliases
PGLYRPIalpha, PGRP-Ialpha, PGRPIA
Chromosome Location
Chr.1: 153297589 - 153312975 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052891.2 NP_443123.1 1 - 2 110 65
XM_011509119.1 XP_011507421.1 2 - 3 431 65
GenBank mRNA AY035376.1 - 1 - 2 110 65
BC069741.1 - 1 - 2 96 65
BC069798.1 - 1 - 2 97 65
BC069801.1 - 1 - 2 97 65
BC128114.1 - 1 - 2 83 65
BC128115.1 - 1 - 2 83 65

View Full Product Details