Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052972.2 | NP_443204.1 | 1 - 2 | 43 | 91 |
GenBank mRNA | AB073607.1 | - | 1 - 2 | 55 | 91 |
AF403428.1 | - | 1 - 2 | 43 | 91 | |
AK027382.1 | - | 1 - 2 | 55 | 91 | |
AK303861.1 | - | 1 - 2 | 55 | 91 | |
AK314506.1 | - | 1 - 2 | 55 | 91 | |
BC034389.1 | - | 1 - 2 | 44 | 91 | |
BC070198.1 | - | 1 - 2 | 100 | 91 |