Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015163.5 | NP_055978.4 | 3 - 4 | 1808 | 79 |
NM_052978.4 | NP_443210.1 | 3 - 4 | 1808 | 79 | |
XM_005267301.3 | XP_005267358.1 | 4 - 5 | 2413 | 79 | |
XM_005267302.3 | XP_005267359.1 | 4 - 5 | 2413 | 79 | |
XM_006720018.3 | XP_006720081.1 | 4 - 5 | 2413 | 79 | |
XM_011536389.2 | XP_011534691.1 | 4 - 5 | 2428 | 79 | |
XM_017020943.1 | XP_016876432.1 | 4 - 5 | 2413 | 79 | |
XM_017020944.1 | XP_016876433.1 | 4 - 5 | 2413 | 79 | |
XM_017020945.1 | XP_016876434.1 | 4 - 5 | 2413 | 79 | |
XM_017020946.1 | XP_016876435.1 | 4 - 5 | 2413 | 79 | |
XM_017020947.1 | XP_016876436.1 | 4 - 5 | 2413 | 79 | |
XM_017020948.1 | XP_016876437.1 | 4 - 5 | 2413 | 79 | |
XM_017020949.1 | XP_016876438.1 | 4 - 5 | 2413 | 79 | |
XM_017020950.1 | XP_016876439.1 | 4 - 5 | 2413 | 79 | |
GenBank mRNA | AK055388.1 | - | 3 - 4 | 1433 | 79 |
BC013414.2 | - | 3 - 4 | 1180 | 79 | |
BC063872.1 | - | 3 - 4 | 2165 | 79 | |
D87458.2 | - | 3 - 4 | 1184 | 79 |