Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080284.2 | NP_525023.2 | 16 - 17 | 2300 | 130 |
XM_011524574.2 | XP_011522876.1 | 10 - 11 | 1628 | 130 | |
XM_017024404.1 | XP_016879893.1 | 16 - 17 | 2612 | 130 | |
XM_017024405.1 | XP_016879894.1 | 16 - 17 | 2612 | 130 | |
XM_017024406.1 | XP_016879895.1 | 12 - 13 | 1580 | 130 | |
XM_017024407.1 | XP_016879896.1 | 13 - 14 | 1684 | 130 | |
XM_017024408.1 | XP_016879897.1 | 2 - 3 | 173 | 130 | |
GenBank mRNA | AF373290.1 | - | 15 - 16 | 2128 | 130 |
AK308431.1 | - | 16 - 17 | 2321 | 131 | |
AY028898.1 | - | 16 - 17 | 2300 | 130 | |
BC125231.1 | - | 16 - 17 | 2299 | 130 | |
BC125232.1 | - | 16 - 17 | 2299 | 130 |