Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307928.1 | NP_001294857.1 | 3 - 4 | 443 | 86 |
NM_080474.2 | NP_536722.1 | 3 - 4 | 383 | 86 | |
XM_005266778.3 | XP_005266835.2 | 4 - 5 | 482 | 86 | |
XM_011526246.1 | XP_011524548.1 | 5 - 6 | 672 | 86 | |
XM_011526248.1 | XP_011524550.1 | 4 - 5 | 508 | 86 | |
XM_011526249.1 | XP_011524551.1 | 4 - 5 | 482 | 86 | |
XM_017026067.1 | XP_016881556.1 | 4 - 5 | 565 | 86 | |
GenBank mRNA | AF411191.1 | - | 3 - 4 | 383 | 86 |
BC103884.1 | - | 3 - 4 | 444 | 86 | |
BC103885.1 | - | 3 - 4 | 443 | 86 |