Assay Design
Gene Transcripts
- Gene Symbol
- DPH3P1
- Entrez Gene ID
-
100132911
- Gene Name
- diphthamide biosynthesis 3 pseudogene 1
- Gene Aliases
-
C20orf143, DPH3B, ZCSL1
- Chromosome Location
-
Chr.20: 62844566 - 62846191 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080750.4 |
NP_542788.1 |
1 - 1 |
1246 |
100 |
- Gene Symbol
- TCFL5
- Entrez Gene ID
-
10732
- Gene Name
- transcription factor like 5
- Gene Aliases
-
CHA, E2BP-1, Figlb, bHLHe82
- Chromosome Location
-
Chr.20: 62841014 - 62861763 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC046933.1 |
- |
6 - 6 |
|
100 |
View Full Product Details