Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318860.1 | NP_001305789.1 | 3 - 4 | 75 | |
NM_001318861.1 | NP_001305790.1 | 4 - 5 | 75 | ||
NM_001318862.1 | NP_001305791.1 | 4 - 5 | 75 | ||
NM_001318863.1 | NP_001305792.1 | 3 - 4 | 75 | ||
NM_080916.2 | NP_550438.1 | 4 - 5 | 75 | ||
XM_011532647.2 | XP_011530949.1 | 4 - 5 | 75 | ||
GenBank mRNA | AI201630.1 | - | 3 - 4 | 482 | 75 |
BC015757.1 | - | 4 - 5 | 75 | ||
BF975904.1 | - | 3 - 4 | 75 | ||
BG470729.1 | - | 3 - 4 | 75 | ||
BG575800.1 | - | 4 - 5 | 75 | ||
CN311954.1 | - | 4 - 5 | 75 | ||
KU178023.1 | - | 4 - 5 | 75 | ||
KU178024.1 | - | 3 - 4 | 75 | ||
U41668.1 | - | 4 - 5 | 75 |