Assay Design



Gene Transcripts

Gene Symbol
HTR3C
Entrez Gene ID
170572
Gene Name
5-hydroxytryptamine receptor 3C
Gene Aliases
-
Chromosome Location
Chr.3: 184053047 - 184060673 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130770.2 NP_570126.2 4 - 5 425 56
GenBank mRNA AF459285.1 - 4 - 5 425 56
BC131799.1 - 4 - 5 416 56

View Full Product Details