Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002149.3 | NP_002140.2 | 1 - 2 | 275 | 72 |
XM_005246161.1 | XP_005246218.1 | 1 - 2 | 290 | 72 | |
XM_005246162.1 | XP_005246219.1 | 1 - 2 | 290 | 72 | |
XM_011510347.1 | XP_011508649.1 | 1 - 2 | 290 | 72 | |
XM_011510349.1 | XP_011508651.1 | 1 - 2 | 290 | 72 | |
GenBank mRNA | BC009846.2 | - | 1 - 2 | 250 | 72 |
BC017028.1 | - | 1 - 2 | 242 | 72 | |
BC017482.1 | - | 1 - 2 | 265 | 72 | |
BI603158.1 | - | 1 - 2 | 280 | 72 |