Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081676.1 | NP_001075145.1 | 1 - 2 | 242 | 78 |
NM_001081677.1 | NP_001075146.1 | 1 - 2 | 242 | 78 | |
NM_006922.3 | NP_008853.3 | 1 - 2 | 242 | 78 | |
XM_011511610.2 | XP_011509912.1 | 1 - 2 | 225 | 78 | |
XM_011511613.2 | XP_011509915.1 | 1 - 2 | 470 | 78 | |
XM_017004660.1 | XP_016860149.1 | 1 - 2 | 225 | 78 | |
XM_017004661.1 | XP_016860150.1 | 1 - 2 | 225 | 78 | |
XM_017004662.1 | XP_016860151.1 | 1 - 2 | 225 | 78 | |
XM_017004663.1 | XP_016860152.1 | 1 - 2 | 225 | 78 | |
XM_017004664.1 | XP_016860153.1 | 1 - 2 | 225 | 78 | |
XM_017004665.1 | XP_016860154.1 | 1 - 2 | 225 | 78 | |
XM_017004666.1 | XP_016860155.1 | 1 - 2 | 225 | 78 | |
GenBank mRNA | AF225986.1 | - | 1 - 2 | 221 | 78 |
AF225987.1 | - | 1 - 2 | 221 | 78 | |
AJ251507.1 | - | 1 - 2 | 61 | 78 | |
AK299757.1 | - | 1 - 2 | 225 | 78 | |
AL079365.1 | - | 1 - 2 | 241 | 78 | |
DA772987.1 | - | 1 - 2 | 225 | 78 | |
DA796981.1 | - | 1 - 2 | 226 | 78 |