Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015072.4 | NP_055887.3 | 22 - 23 | 2559 | 96 |
GenBank mRNA | AB023215.1 | - | 20 - 21 | 2119 | 96 |
AK309748.1 | - | 23 - 24 | 2541 | 96 | |
AL136808.1 | - | 8 - 9 | 1007 | 96 | |
AY237126.1 | - | 22 - 23 | 2489 | 96 | |
BC016625.1 | - | 9 - 10 | 1140 | 96 | |
BC136472.1 | - | 22 - 23 | 2449 | 96 | |
BC136473.1 | - | 22 - 23 | 2449 | 96 | |
CR533554.1 | - | 7 - 8 | 937 | 96 |