Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015230.3 | NP_056045.2 | 2 - 3 | 1244 | 99 |
XM_017007698.1 | XP_016863187.1 | 2 - 3 | 1674 | 99 | |
XM_017007699.1 | XP_016863188.1 | 2 - 3 | 1129 | 99 | |
XM_017007700.1 | XP_016863189.1 | 2 - 3 | 1674 | 99 | |
XM_017007701.1 | XP_016863190.1 | 2 - 3 | 1674 | 99 | |
XM_017007702.1 | XP_016863191.1 | 2 - 3 | 1674 | 99 | |
GenBank mRNA | AB011152.2 | - | 1 - 2 | 687 | 99 |
AF439781.1 | - | 1 - 2 | 1066 | 99 | |
AK126864.1 | - | 2 - 3 | 1237 | 99 | |
AY049733.1 | - | 2 - 3 | 1159 | 99 | |
BC150258.1 | - | 1 - 2 | 698 | 99 |