Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025604.2 | NP_001020775.1 | 2 - 3 | 512 | 61 |
NM_001286826.1 | NP_001273755.1 | 2 - 3 | 487 | 61 | |
NM_015683.1 | NP_056498.1 | 2 - 3 | 487 | 61 | |
GenBank mRNA | AF193051.1 | - | 2 - 3 | 487 | 61 |
AK127940.1 | - | 2 - 3 | 476 | 61 | |
AK314657.1 | - | 2 - 3 | 485 | 61 | |
BC022516.1 | - | 2 - 3 | 485 | 61 | |
BG756314.1 | - | 2 - 3 | 513 | 61 |