Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 16 - 17 | 1748 | 81 |
NM_020320.4 | NP_064716.2 | 17 - 18 | 1583 | 81 | |
NR_134857.1 | 17 - 18 | 1579 | 81 | ||
XM_011535948.2 | XP_011534250.1 | 17 - 18 | 1568 | 81 | |
XM_017011072.1 | XP_016866561.1 | 17 - 18 | 1749 | 81 | |
XM_017011073.1 | XP_016866562.1 | 18 - 19 | 1967 | 81 | |
XM_017011074.1 | XP_016866563.1 | 17 - 18 | 1570 | 81 | |
XM_017011075.1 | XP_016866564.1 | 16 - 17 | 1693 | 81 | |
XM_017011076.1 | XP_016866565.1 | 18 - 19 | 1911 | 81 | |
XM_017011077.1 | XP_016866566.1 | 16 - 17 | 1470 | 81 | |
XM_017011078.1 | XP_016866567.1 | 16 - 17 | 1514 | 81 | |
XM_017011079.1 | XP_016866568.1 | 17 - 18 | 1749 | 81 | |
XM_017011080.1 | XP_016866569.1 | 18 - 19 | 1911 | 81 | |
XM_017011081.1 | XP_016866570.1 | 16 - 17 | 1514 | 81 | |
GenBank mRNA | AK023550.1 | - | 17 - 18 | 1526 | 81 |
AK093934.1 | - | 17 - 18 | 1956 | 81 | |
AK315669.1 | - | 17 - 18 | 1520 | 81 | |
BC010420.1 | - | 17 - 18 | 1554 | 81 | |
BC105784.1 | - | 16 - 17 | 1687 | 81 | |
CA415020.1 | - | 3 - 4 | 81 |