Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003712.1 | NP_001003712.1 | 18 - 19 | 2483 | 119 |
NM_001319652.1 | NP_001306581.1 | 21 - 22 | 2608 | 119 | |
NM_001319653.1 | NP_001306582.1 | 19 - 20 | 2520 | 119 | |
NM_001319655.1 | NP_001306584.1 | 16 - 17 | 2414 | 119 | |
NM_020841.4 | NP_065892.1 | 19 - 20 | 2529 | 119 | |
XM_005268621.4 | XP_005268678.1 | 22 - 23 | 2627 | 119 | |
XM_006719224.2 | XP_006719287.1 | 18 - 19 | 2488 | 119 | |
XM_011537855.2 | XP_011536157.1 | 19 - 20 | 2146 | 119 | |
XM_011537856.2 | XP_011536158.1 | 20 - 21 | 2564 | 119 | |
XM_011537857.2 | XP_011536159.1 | 22 - 23 | 2618 | 119 | |
XM_017018768.1 | XP_016874257.1 | 19 - 20 | 2161 | 119 | |
XM_017018769.1 | XP_016874258.1 | 20 - 21 | 2532 | 119 | |
GenBank mRNA | AB040884.3 | - | 19 - 20 | 2200 | 119 |
AF392452.1 | - | 19 - 20 | 2530 | 119 | |
AK092160.1 | - | 16 - 17 | 2415 | 119 | |
AK289997.1 | - | 19 - 20 | 2249 | 119 | |
AL832876.1 | - | 6 - 7 | 585 | 119 | |
BC012935.1 | - | 5 - 6 | 514 | 119 | |
BC082235.1 | - | 19 - 20 | 2468 | 119 | |
BC093834.1 | - | 18 - 19 | 2128 | 119 | |
BC101529.1 | - | 18 - 19 | 2128 | 119 | |
BC111728.1 | - | 19 - 20 | 2423 | 119 | |
BX647211.1 | - | 18 - 19 | 2085 | 119 | |
CR749542.1 | - | 21 - 22 | 2333 | 119 |