Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021044.3 | NP_066382.1 | 2 - 3 | 872 | 98 |
XM_017019380.1 | XP_016874869.1 | 2 - 3 | 803 | 98 | |
XM_017019381.1 | XP_016874870.1 | 2 - 3 | 520 | 98 | |
GenBank mRNA | AK313360.1 | - | 2 - 3 | 842 | 98 |
BC033507.1 | - | 2 - 3 | 872 | 98 | |
HQ607677.1 | - | 2 - 3 | 869 | 98 | |
HQ607678.1 | - | 2 - 3 | 876 | 98 |