Assay Design



Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
58484
Gene Name
NLR family CARD domain containing 4
Gene Aliases
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
Chromosome Location
Chr.2: 32224449 - 32265743 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199138.1 NP_001186067.1 2 - 3 263 108
NM_001199139.1 NP_001186068.1 2 - 3 146 108
NM_001302504.1 NP_001289433.1 2 - 3 277 108
NM_021209.4 NP_067032.3 2 - 3 277 108
XM_017004619.1 XP_016860108.1 2 - 3 263 108
GenBank mRNA AF376061.1 - 3 - 4 491 108
AK095467.1 - 2 - 3 263 108
AK292673.1 - 2 - 3 248 108
AK314762.1 - 2 - 3 248 108
AY027787.1 - 2 - 3 278 108
AY027788.1 - 2 - 3 278 108
AY027789.1 - 2 - 3 278 108
AY027790.1 - 2 - 3 278 108
AY035391.1 - 2 - 3 146 108
AY358152.1 - 2 - 3 225 108
BC031555.1 - 2 - 3 233 108
DN998613.1 - 2 - 3 283 108

View Full Product Details