Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033046.2 | NP_149035.1 | 2 - 3 | 658 | 79 |
XM_017004635.1 | XP_016860124.1 | 1 - 2 | 768 | 79 | |
XM_017004636.1 | XP_016860125.1 | 2 - 3 | 662 | 79 | |
GenBank mRNA | AK307302.1 | - | 2 - 3 | 528 | 79 |
BC017727.1 | - | 1 - 2 | 270 | 79 | |
BQ721791.1 | - | 2 - 3 | 600 | 79 |