Assay Design



Gene Transcripts

Gene Symbol
MGME1
Entrez Gene ID
92667
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, DDK1, MTDPS11, bA504H3.4
Chromosome Location
Chr.20: 17968594 - 17991122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001310338.1 NP_001297267.1 4 - 5 1086 121
NM_052865.3 NP_443097.1 3 - 4 1041 121
XM_005260867.2 XP_005260924.1 4 - 5 1308 121
XM_011529394.1 XP_011527696.1 4 - 5 923 121
XM_011529395.1 XP_011527697.1 3 - 4 801 121
XM_017028127.1 XP_016883616.1 3 - 4 1263 121
XM_017028128.1 XP_016883617.1 3 - 4 1137 121
GenBank mRNA AF306684.1 - 3 - 4 850 121
AF306685.1 - 3 - 4 850 121
AK027503.1 - 3 - 4 845 121
AK310282.1 - 2 - 3 357 121
AK315532.1 - 3 - 4 877 121
BC016869.1 - 3 - 4 813 121

View Full Product Details