Assay Design



Gene Transcripts

Gene Symbol
ACSM1
Entrez Gene ID
116285
Gene Name
acyl-CoA synthetase medium-chain family member 1
Gene Aliases
BUCS1, MACS1
Chromosome Location
Chr.16: 20623235 - 20697788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318890.1 NP_001305819.1 7 - 8 1198 74
NM_052956.2 NP_443188.2 6 - 7 1062 74
XM_006721016.2 XP_006721079.1 6 - 7 1041 74
XM_006721017.2 XP_006721080.1 6 - 7 1041 74
XM_006721018.2 XP_006721081.1 4 - 5 420 74
XM_011545729.2 XP_011544031.1 6 - 7 1041 74
XM_011545731.2 XP_011544033.1 4 - 5 1237 74
XM_011545732.2 XP_011544034.1 5 - 6 479 74
XM_017022915.1 XP_016878404.1 5 - 6 1889 74
GenBank mRNA AB059429.1 - 6 - 7 1062 74
AK092574.1 - 4 - 5 420 74
AK314282.1 - 7 - 8 1154 74
BC125178.1 - 6 - 7 1062 74

View Full Product Details