Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193476.1 | NP_001180405.1 | 11 - 12 | 1479 | 83 |
NM_052961.3 | NP_443193.1 | 11 - 12 | 1503 | 83 | |
NM_138718.2 | NP_619732.2 | 9 - 10 | 1226 | 83 | |
XM_017010235.1 | XP_016865724.1 | 11 - 12 | 1529 | 83 | |
GenBank mRNA | AF314959.1 | - | 10 - 11 | 1373 | 83 |
AF331522.1 | - | 11 - 12 | 1447 | 83 | |
AK055314.1 | - | 1 - 2 | 949 | 83 | |
AK097410.1 | - | 5 - 6 | 565 | 83 | |
AK122928.1 | - | 11 - 12 | 1479 | 83 | |
AK125189.1 | - | 11 - 12 | 1723 | 83 | |
BC025408.2 | - | 9 - 10 | 1219 | 83 |