Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054023.4 | NP_473364.1 | 1 - 2 | 145 | 82 |
GenBank mRNA | AF313455.1 | - | 1 - 2 | 66 | 82 |
AF439545.1 | - | 1 - 2 | 134 | 82 | |
AY358979.1 | - | 1 - 2 | 134 | 82 | |
BC024232.1 | - | 1 - 2 | 149 | 82 |