Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080476.4 | NP_536724.1 | 4 - 5 | 516 | 83 |
XM_017027664.1 | XP_016883153.1 | 4 - 5 | 334 | 83 | |
XM_017027665.1 | XP_016883154.1 | 4 - 5 | 334 | 83 | |
GenBank mRNA | AB086842.1 | - | 4 - 5 | 322 | 83 |
AK075507.1 | - | 4 - 5 | 341 | 83 | |
AK301900.1 | - | 4 - 5 | 316 | 83 | |
AK308627.1 | - | 3 - 4 | 263 | 83 | |
AY339061.1 | - | 3 - 4 | 256 | 83 | |
AY358816.1 | - | 4 - 5 | 346 | 83 | |
AY422169.1 | - | 4 - 5 | 516 | 83 | |
BC030512.1 | - | 4 - 5 | 327 | 83 |