Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316676.1 | NP_001303605.1 | 12 - 13 | 1293 | 87 |
NM_001316677.1 | NP_001303606.1 | 10 - 11 | 966 | 87 | |
NM_001323354.1 | NP_001310283.1 | 10 - 11 | 2686 | 87 | |
NM_001323355.1 | NP_001310284.1 | 10 - 11 | 1099 | 87 | |
NM_001323356.1 | NP_001310285.1 | 10 - 11 | 1093 | 87 | |
NM_001323357.1 | NP_001310286.1 | 7 - 8 | 678 | 87 | |
NM_006504.5 | NP_006495.1 | 11 - 12 | 1122 | 87 | |
NM_130435.4 | NP_569119.1 | 8 - 9 | 769 | 87 | |
XM_005252691.2 | XP_005252748.1 | 10 - 11 | 966 | 87 | |
XM_011539995.1 | XP_011538297.1 | 10 - 11 | 1084 | 87 | |
XM_011539996.1 | XP_011538298.1 | 10 - 11 | 1084 | 87 | |
XM_011539998.1 | XP_011538300.1 | 10 - 11 | 1084 | 87 | |
XM_017016467.1 | XP_016871956.1 | 10 - 11 | 1084 | 87 | |
XM_017016468.1 | XP_016871957.1 | 10 - 11 | 873 | 87 | |
XM_017016469.1 | XP_016871958.1 | 10 - 11 | 1084 | 87 | |
GenBank mRNA | AJ315969.1 | - | 8 - 9 | 678 | 87 |
AJ430580.1 | - | 8 - 9 | 687 | 87 | |
AK290414.1 | - | 8 - 9 | 705 | 87 | |
AK291828.1 | - | 10 - 11 | 966 | 87 | |
AK299498.1 | - | 9 - 10 | 844 | 87 | |
AK300613.1 | - | 9 - 10 | 859 | 87 | |
BC050062.1 | - | 12 - 13 | 1201 | 87 | |
BX648180.1 | - | 7 - 8 | 684 | 87 | |
X54134.1 | - | 11 - 12 | 894 | 87 |