Assay Design



Gene Transcripts

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location
Chr.19: 5205503 - 5340803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002850.3 NP_002841.3 2 - 3 326 72
NM_130853.2 NP_570923.2 2 - 3 326 72
NM_130854.2 NP_570924.2 2 - 3 326 72
NM_130855.2 NP_570925.2 2 - 3 326 72
XM_005259600.2 XP_005259657.1 2 - 3 294 72
XM_005259606.2 XP_005259663.1 2 - 3 294 72
XM_005259607.2 XP_005259664.1 2 - 3 294 72
XM_005259609.1 XP_005259666.1 2 - 3 293 72
XM_005259610.1 XP_005259667.1 2 - 3 293 72
XM_011528157.2 XP_011526459.1 1 - 2 164 72
XM_017027065.1 XP_016882554.1 2 - 3 294 72
XM_017027066.1 XP_016882555.1 2 - 3 312 72
XM_017027067.1 XP_016882556.1 2 - 3 210 72
XM_017027068.1 XP_016882557.1 2 - 3 294 72
XM_017027069.1 XP_016882558.1 2 - 3 294 72
XM_017027070.1 XP_016882559.1 2 - 3 294 72
XM_017027071.1 XP_016882560.1 2 - 3 294 72
XM_017027072.1 XP_016882561.1 2 - 3 294 72
XM_017027074.1 XP_016882563.1 2 - 3 293 72
XM_017027075.1 XP_016882564.1 2 - 3 293 72
XM_017027076.1 XP_016882565.1 2 - 3 293 72
GenBank mRNA AB209333.1 - 2 - 3 326 72
BC029496.1 - 1 - 2 340 72
BC104812.1 - 1 - 2 166 72
BC143286.1 - 1 - 2 166 72
BC143287.1 - 1 - 2 166 72
BI461316.1 - 1 - 2 345 72
U35234.1 - 1 - 2 200 72
U40317.1 - 1 - 2 114 72
U41725.1 - 1 - 2 114 72

View Full Product Details