Assay Design



Gene Transcripts

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
Chromosome Location
Chr.19: 5205503 - 5340803 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002850.3 NP_002841.3 4 - 5 617 76
NM_130853.2 NP_570923.2 4 - 5 617 76
NM_130854.2 NP_570924.2 4 - 5 617 76
NM_130855.2 NP_570925.2 4 - 5 617 76
XM_005259600.2 XP_005259657.1 4 - 5 585 76
XM_005259606.2 XP_005259663.1 4 - 5 585 76
XM_005259607.2 XP_005259664.1 4 - 5 585 76
XM_005259609.1 XP_005259666.1 4 - 5 584 76
XM_005259610.1 XP_005259667.1 4 - 5 584 76
XM_011528157.2 XP_011526459.1 3 - 4 455 76
XM_017027065.1 XP_016882554.1 4 - 5 585 76
XM_017027066.1 XP_016882555.1 4 - 5 603 76
XM_017027067.1 XP_016882556.1 4 - 5 501 76
XM_017027068.1 XP_016882557.1 4 - 5 585 76
XM_017027069.1 XP_016882558.1 4 - 5 585 76
XM_017027070.1 XP_016882559.1 4 - 5 585 76
XM_017027071.1 XP_016882560.1 4 - 5 585 76
XM_017027072.1 XP_016882561.1 4 - 5 585 76
XM_017027074.1 XP_016882563.1 4 - 5 584 76
XM_017027075.1 XP_016882564.1 4 - 5 584 76
XM_017027076.1 XP_016882565.1 4 - 5 584 76
GenBank mRNA AB209333.1 - 4 - 5 617 76
BC104812.1 - 3 - 4 457 76
BC143286.1 - 3 - 4 457 76
BC143287.1 - 3 - 4 457 76
U35234.1 - 3 - 4 491 76
U40317.1 - 3 - 4 405 76
U41725.1 - 3 - 4 405 76

View Full Product Details