Assay Design



Gene Transcripts

Gene Symbol
NETO1
Entrez Gene ID
81832
Gene Name
neuropilin and tolloid like 1
Gene Aliases
BCTL1, BTCL1
Chromosome Location
Chr.18: 72742314 - 72867667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201465.1 NP_001188394.1 6 - 7 922 78
NM_138966.3 NP_620416.1 6 - 7 922 78
XM_005266774.4 XP_005266831.1 6 - 7 860 78
XM_017026017.1 XP_016881506.1 6 - 7 784 78
XM_017026018.1 XP_016881507.1 6 - 7 784 78
XM_017026019.1 XP_016881508.1 6 - 7 944 78
XM_017026020.1 XP_016881509.1 6 - 7 948 78
XM_017026021.1 XP_016881510.1 6 - 7 762 78
GenBank mRNA AF448838.1 - 6 - 7 928 78
AL834354.1 - 4 - 5 541 78
BC050329.1 - 6 - 7 924 78

View Full Product Details