Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201465.1 | NP_001188394.1 | 8 - 9 | 1260 | 116 |
NM_138966.3 | NP_620416.1 | 8 - 9 | 1260 | 116 | |
XM_005266774.4 | XP_005266831.1 | 8 - 9 | 1198 | 116 | |
XM_017026017.1 | XP_016881506.1 | 8 - 9 | 1122 | 116 | |
XM_017026018.1 | XP_016881507.1 | 8 - 9 | 1122 | 116 | |
XM_017026019.1 | XP_016881508.1 | 8 - 9 | 1282 | 116 | |
XM_017026020.1 | XP_016881509.1 | 8 - 9 | 1286 | 116 | |
XM_017026021.1 | XP_016881510.1 | 8 - 9 | 1100 | 116 | |
GenBank mRNA | AF448838.1 | - | 8 - 9 | 1266 | 116 |
AL834354.1 | - | 6 - 7 | 879 | 116 | |
BC050329.1 | - | 8 - 9 | 1262 | 116 |