Assay Design
Gene Transcripts
- Gene Symbol
- DCHS1
- Entrez Gene ID
-
8642
- Gene Name
- dachsous cadherin-related 1
- Gene Aliases
-
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
- Chromosome Location
-
Chr.11: 6621324 - 6655849 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_003737.3 |
NP_003728.1 |
9 - 10 |
4337 |
93 |
GenBank mRNA |
AB053446.1 |
- |
9 - 10 |
4340 |
93 |
View Full Product Details