Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024383.1 | NP_001019554.1 | 17 - 18 | 4450 | 68 |
NM_014903.5 | NP_055718.4 | 17 - 18 | 4450 | 68 | |
XM_005269215.3 | XP_005269272.1 | 17 - 18 | 8451 | 68 | |
XM_017020164.1 | XP_016875653.1 | 17 - 18 | 8453 | 68 | |
XM_017020166.1 | XP_016875655.1 | 17 - 18 | 4439 | 68 | |
XM_017020167.1 | XP_016875656.1 | 16 - 17 | 4370 | 68 | |
XM_017020168.1 | XP_016875657.1 | 16 - 17 | 7963 | 68 | |
XM_017020171.1 | XP_016875660.1 | 9 - 10 | 7746 | 68 | |
XM_017020173.1 | XP_016875662.1 | 8 - 9 | 7255 | 68 | |
XM_017020175.1 | XP_016875664.1 | 8 - 9 | 3134 | 68 | |
GenBank mRNA | AF397731.1 | - | 16 - 17 | 4034 | 68 |
AJ488201.1 | - | 17 - 18 | 4322 | 68 |