Assay Design



Gene Transcripts

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
Chromosome Location
Chr.12: 77571862 - 78213012 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024383.1 NP_001019554.1 21 - 22 4879 80
NM_014903.5 NP_055718.4 21 - 22 4879 80
XM_005269215.3 XP_005269272.1 21 - 22 8880 80
XM_011538940.2 XP_011537242.1 20 - 21 8839 80
XM_011538941.2 XP_011537243.1 20 - 21 8838 80
XM_011538944.2 XP_011537246.1 19 - 20 8347 80
XM_017020164.1 XP_016875653.1 21 - 22 8882 80
XM_017020165.1 XP_016875654.1 20 - 21 8840 80
XM_017020166.1 XP_016875655.1 21 - 22 4868 80
XM_017020167.1 XP_016875656.1 20 - 21 4799 80
XM_017020168.1 XP_016875657.1 20 - 21 8392 80
XM_017020169.1 XP_016875658.1 19 - 20 8349 80
XM_017020170.1 XP_016875659.1 19 - 20 8346 80
XM_017020171.1 XP_016875660.1 13 - 14 8175 80
XM_017020172.1 XP_016875661.1 12 - 13 3771 80
XM_017020173.1 XP_016875662.1 12 - 13 7684 80
XM_017020174.1 XP_016875663.1 11 - 12 7642 80
XM_017020175.1 XP_016875664.1 12 - 13 3563 80
GenBank mRNA AB023155.2 - 6 - 7 1389 80
AF397731.1 - 20 - 21 4463 80
AJ488201.1 - 21 - 22 4751 80
AK298525.1 - 5 - 6 593 80

View Full Product Details