Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195139.1 | NP_001182068.1 | 1 - 2 | 186 | 94 |
NM_015386.2 | NP_056201.2 | 1 - 2 | 186 | 94 | |
XM_011522981.2 | XP_011521283.1 | 1 - 2 | 173 | 94 | |
GenBank mRNA | AB088369.1 | - | 1 - 2 | 163 | 94 |
AK022874.1 | - | 1 - 2 | 166 | 94 | |
AK096557.1 | - | 1 - 2 | 186 | 94 | |
AK293235.1 | - | 1 - 2 | 184 | 94 | |
AK297557.1 | - | 2 - 3 | 129 | 94 | |
BC027726.2 | - | 1 - 2 | 160 | 94 | |
BC072438.1 | - | 1 - 2 | 166 | 94 | |
BC128144.1 | - | 1 - 2 | 163 | 94 | |
BC128145.1 | - | 1 - 2 | 163 | 94 |