Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284377.1 | NP_001271306.1 | 3 - 4 | 232 | 75 |
NM_001284378.1 | NP_001271307.1 | 3 - 4 | 232 | 75 | |
NM_001284379.1 | NP_001271308.1 | 2 - 3 | 210 | 75 | |
NM_001321844.1 | NP_001308773.1 | 3 - 4 | 226 | 75 | |
NM_001321848.1 | NP_001308777.1 | 2 - 3 | 210 | 75 | |
NM_017828.4 | NP_060298.2 | 3 - 4 | 232 | 75 | |
NR_104312.1 | 3 - 4 | 232 | 75 | ||
NR_104313.1 | 3 - 4 | 232 | 75 | ||
XM_005254509.3 | XP_005254566.1 | 3 - 4 | 258 | 75 | |
XM_011521739.1 | XP_011520041.1 | 3 - 4 | 267 | 75 | |
XM_011521741.1 | XP_011520043.1 | 3 - 4 | 267 | 75 | |
GenBank mRNA | AK000459.1 | - | 3 - 4 | 169 | 75 |
AK225716.1 | - | 3 - 4 | 193 | 75 | |
AK293708.1 | - | 3 - 4 | 168 | 75 | |
AK314740.1 | - | 3 - 4 | 165 | 75 | |
AL136872.1 | - | 3 - 4 | 152 | 75 | |
AY542160.1 | - | 2 - 3 | 138 | 75 | |
BC000837.2 | - | 2 - 3 | 135 | 75 | |
BC068998.1 | - | 3 - 4 | 154 | 75 | |
BI829264.1 | - | 3 - 4 | 208 | 75 | |
BI829484.1 | - | 3 - 4 | 232 | 75 | |
BQ233598.1 | - | - | 195 | 75 | |
BU540913.1 | - | 3 - 4 | 174 | 75 | |
CR533452.1 | - | 2 - 3 | 138 | 75 | |
HY162014.1 | - | 2 - 3 | 211 | 75 |