Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018206.5 | NP_060676.2 | 12 - 13 | 1623 | 72 |
XM_005256045.3 | XP_005256102.1 | 11 - 12 | 1508 | 72 | |
XM_011523227.2 | XP_011521529.1 | 12 - 13 | 2353 | 72 | |
GenBank mRNA | AF175265.1 | - | 12 - 13 | 1581 | 72 |
AF183418.1 | - | 11 - 12 | 1532 | 72 | |
AF186382.1 | - | 12 - 13 | 1577 | 72 | |
AF191298.2 | - | 12 - 13 | 1551 | 72 | |
AK001614.1 | - | 12 - 13 | 1571 | 72 | |
AK023650.1 | - | 8 - 9 | 1134 | 72 | |
AK025774.1 | - | - | 3231 | 72 | |
AL136888.1 | - | 12 - 13 | 1571 | 72 | |
AL512769.1 | - | 5 - 6 | 628 | 72 | |
AY007112.1 | - | 1 - 2 | 134 | 72 | |
BC002414.2 | - | 11 - 12 | 1530 | 72 | |
BC010362.1 | - | 12 - 13 | 1544 | 72 | |
BC093036.1 | - | 12 - 13 | 1569 | 72 | |
BX647213.1 | - | 7 - 8 | 1913 | 72 |