Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033566.2 | NP_001028738.1 | 12 - 13 | 1190 | 119 |
NM_001033567.2 | NP_001028739.2 | 13 - 14 | 1314 | 119 | |
NM_001033568.2 | NP_001028740.1 | 12 - 13 | 1190 | 119 | |
NM_001288754.1 | NP_001275683.1 | 12 - 13 | 1190 | 119 | |
NM_001288755.1 | NP_001275684.1 | 12 - 13 | 1186 | 119 | |
NM_001288758.1 | NP_001275687.1 | 13 - 14 | 1314 | 119 | |
NM_018307.4 | NP_060777.3 | 12 - 13 | 1190 | 119 | |
NR_110083.1 | 12 - 13 | 1190 | 119 | ||
XM_011524969.2 | XP_011523271.1 | 11 - 12 | 914 | 119 | |
XM_011524971.2 | XP_011523273.2 | 12 - 13 | 986 | 119 | |
XM_011524973.2 | XP_011523275.2 | 12 - 13 | 986 | 119 | |
XM_011524975.2 | XP_011523277.1 | 13 - 14 | 1443 | 119 | |
XM_011524976.2 | XP_011523278.2 | 12 - 13 | 986 | 119 | |
XM_017024819.1 | XP_016880308.1 | 13 - 14 | 1218 | 119 | |
XM_017024820.1 | XP_016880309.1 | 11 - 12 | 914 | 119 | |
XM_017024821.1 | XP_016880310.1 | 12 - 13 | 986 | 119 | |
XM_017024822.1 | XP_016880311.1 | 12 - 13 | 1215 | 119 | |
XM_017024823.1 | XP_016880312.1 | 12 - 13 | 986 | 119 | |
XM_017024824.1 | XP_016880313.1 | 13 - 14 | 1321 | 119 | |
XM_017024825.1 | XP_016880314.1 | 13 - 14 | 1413 | 119 | |
GenBank mRNA | AJ496730.1 | - | 12 - 13 | 1178 | 119 |
AJ517412.1 | - | 12 - 13 | 951 | 119 | |
AK001902.1 | - | 6 - 7 | 554 | 119 | |
AK022695.1 | - | 5 - 6 | 484 | 119 | |
AK294407.1 | - | 12 - 13 | 964 | 119 | |
AL136929.1 | - | 12 - 13 | 1014 | 119 | |
AY094972.1 | - | 12 - 13 | 1067 | 119 | |
BC041114.1 | - | 12 - 13 | 1124 | 119 | |
BC051818.1 | - | 11 - 12 | 901 | 119 | |
BC060781.1 | - | 12 - 13 | 1190 | 119 | |
BC068463.1 | - | 12 - 13 | 1078 | 119 | |
BC125104.1 | - | 12 - 13 | 1019 | 119 | |
BC125105.1 | - | 12 - 13 | 1019 | 119 |