Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017013587.1 | XP_016869076.1 | 1 - 2 | 243 | 70 |
GenBank mRNA | AF155661.1 | - | 2 - 3 | 198 | 70 |
AK302651.1 | - | 1 - 2 | 114 | 70 | |
DC369809.1 | - | 1 - 2 | 185 | 70 |