Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199196.1 | NP_001186125.1 | 5 - 6 | 1284 | 60 |
NM_033415.3 | NP_219483.1 | 4 - 5 | 1176 | 60 | |
XM_005260157.3 | XP_005260214.1 | 4 - 5 | 1227 | 60 | |
XM_017027484.1 | XP_016882973.1 | 4 - 5 | 1151 | 60 | |
XM_017027485.1 | XP_016882974.1 | 6 - 7 | 1408 | 60 | |
GenBank mRNA | AK295488.1 | - | 4 - 5 | 950 | 60 |
AK303914.1 | - | 2 - 3 | 864 | 60 | |
BC056161.1 | - | 6 - 7 | 906 | 60 | |
BC067125.1 | - | 5 - 6 | 1053 | 60 | |
BX648486.1 | - | 4 - 5 | 1071 | 60 | |
CR749678.1 | - | 1 - 2 | 517 | 60 |