Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057749.2 | NP_477097.1 | 1 - 2 | 81 | 70 |
GenBank mRNA | BC007015.1 | - | 1 - 2 | 28 | 70 |
BC020729.1 | - | 1 - 2 | 28 | 70 | |
BG720611.1 | - | 1 - 2 | 86 | 70 | |
BJ990268.1 | - | 1 - 2 | 81 | 70 |