Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_078471.3 | NP_510880.2 | 19 - 20 | 3491 | 71 |
NM_203318.1 | NP_976063.1 | 19 - 20 | 3491 | 71 | |
GenBank mRNA | AB177858.1 | - | 18 - 19 | 3310 | 71 |
AB177860.1 | - | 19 - 20 | 2443 | 71 | |
AK302505.1 | - | 18 - 19 | 2244 | 71 | |
AK302725.1 | - | 3 - 4 | 205 | 71 | |
AK303522.1 | - | 18 - 19 | 2381 | 71 | |
AY703984.1 | - | 19 - 20 | 2443 | 71 | |
BC039612.1 | - | 19 - 20 | 3595 | 71 | |
CR933614.1 | - | 19 - 20 | 3466 | 71 | |
D86970.2 | - | 19 - 20 | 3514 | 71 |