Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308264.1 | NP_001295193.1 | 2 - 3 | 444 | 90 |
NM_017744.4 | NP_060214.2 | 2 - 3 | 598 | 90 | |
NM_138728.2 | NP_620056.1 | 2 - 3 | 598 | 90 | |
NM_138729.3 | NP_620057.1 | 2 - 3 | 598 | 90 | |
XM_005270963.4 | XP_005271020.1 | 2 - 3 | 121 | 90 | |
XM_011541627.2 | XP_011539929.1 | 2 - 3 | 307 | 90 | |
XM_011541628.2 | XP_011539930.1 | 2 - 3 | 307 | 90 | |
XM_011541629.2 | XP_011539931.1 | 2 - 3 | 321 | 90 | |
XM_011541630.2 | XP_011539932.1 | 2 - 3 | 1017 | 90 | |
XM_011541631.2 | XP_011539933.1 | 2 - 3 | 145 | 90 | |
XM_017001534.1 | XP_016857023.1 | 2 - 3 | 307 | 90 | |
XM_017001535.1 | XP_016857024.1 | 2 - 3 | 307 | 90 | |
XM_017001537.1 | XP_016857026.1 | 1 - 2 | 88 | 90 | |
XM_017001538.1 | XP_016857027.1 | 1 - 2 | 88 | 90 | |
GenBank mRNA | AB081317.1 | - | 2 - 3 | 369 | 90 |
AB081319.1 | - | 2 - 3 | 369 | 90 | |
AB081320.1 | - | 2 - 3 | 369 | 90 | |
AK291042.1 | - | 2 - 3 | 299 | 90 | |
AK295507.1 | - | 2 - 3 | 296 | 90 | |
AK295955.1 | - | 2 - 3 | 296 | 90 | |
AK301869.1 | - | 2 - 3 | 299 | 90 | |
BC037830.1 | - | 2 - 3 | 1011 | 90 | |
BC047392.1 | - | 2 - 3 | 324 | 90 | |
BC065501.1 | - | 2 - 3 | 505 | 90 | |
BX537988.1 | - | 3 - 4 | 460 | 90 | |
CD358134.1 | - | 2 - 3 | 598 | 90 |