Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138801.2 | NP_620156.1 | 2 - 3 | 596 | 78 |
XM_011532540.1 | XP_011530842.1 | 2 - 3 | 596 | 78 | |
GenBank mRNA | AK291489.1 | - | 2 - 3 | 404 | 78 |
BC014916.1 | - | 2 - 3 | 378 | 78 | |
BC019263.1 | - | 2 - 3 | 409 | 78 | |
EU794611.1 | - | 2 - 3 | 378 | 78 |