Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139284.2 | NP_644813.1 | 6 - 7 | 1218 | 60 |
XM_011526595.1 | XP_011524897.1 | 7 - 8 | 699 | 60 | |
XM_017026428.1 | XP_016881917.1 | 6 - 7 | 547 | 60 | |
XM_017026429.1 | XP_016881918.1 | 7 - 8 | 619 | 60 | |
XM_017026430.1 | XP_016881919.1 | 5 - 6 | 401 | 60 | |
GenBank mRNA | AF467954.1 | - | 6 - 7 | 910 | 60 |
AJ487519.1 | - | 6 - 7 | 1143 | 60 | |
AJ487959.1 | - | 6 - 7 | 651 | 60 | |
AL832968.1 | - | 4 - 5 | 359 | 60 | |
BC029943.1 | - | 7 - 8 | 683 | 60 | |
BC136694.1 | - | 6 - 7 | 693 | 60 | |
BC136697.1 | - | 6 - 7 | 693 | 60 |